Bexarotene ≥99%

Supplier: Thermo Scientific Chemicals
Warning

J63701-EXH J63701-MC
CAAAJ63701-EXHEA 965.03 CAD
CAAAJ63701-EXH CAAAJ63701-MC
Bexarotene ≥99%
Bexarotene

Synthetic retinoid analog with specific affinity for the retinoid X receptor


Powder


Synthetic retinoid analog with specific affinity for the retinoid X receptor

Formula: C₂₄H₂₈O₂
Melting Pt: 220…225 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00932428
CAS Number: 153559-49-0
Merck Index: 14,1194

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR