Neocuproine 98%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 2,9-Dimethyl-1,10-phenanthroline

J63698-06 J63698-14
CAAAJ63698-06EA 128.28 CAD
CAAAJ63698-06 CAAAJ63698-14
Neocuproine 98%
Neocuproine

Reagent for the colorimetric determination of copper


Powder


Reagent for the colorimetric determination of copper

Formula: C₁₄H₁₂N₂
MW: 208.26 g/mol
Melting Pt: 159…164 °C
Storage Temperature: Ambient
MDL Number: MFCD00004973
CAS Number: 484-11-7
EINECS: 207-601-9
Merck Index: 14,6449

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR