TRIS-buffered saline (TBS, 20X), with 0.5% Tween 20, Thermo Scientific
Supplier: Thermo Scientific Chemicals
J63682-K2
J63682-K3
CAAAJ63682-K2EA
161.79
CAD
CAAAJ63682-K2
CAAAJ63682-K3
TRIS-buffered saline (TBS, 20X), with 0.5% Tween 20, Thermo Scientific
Buffers
Electrophoresis Buffers
Washing Buffers
Liquid
Physical Form: Liquid
MDL No.: MFCD00678198
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...