Domperidone

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 5-Chloro-1-[1-[3-(2-oxo-1-benzimidazolyl)propyl]-4-piperidyl]benzimidazol-2-one

J63681-MC J63681-ME
CAAAJ63681-MCEA 317.98 CAD
CAAAJ63681-MC CAAAJ63681-ME
Domperidone
Domperidone

Peripheral dopamine receptor antagonist that does not cross the blood-brain barrier


Crystalline powder


Applications: Peripheral dopamine receptor antagonist that does not cross the blood-brain barrier

Formula: C₂₂H₂₄ClN₅O₂
Melting Pt: 243 °C
Storage Temperature: Ambient
MDL Number: MFCD00069256
CAS Number: 57808-66-9
EINECS: 260-968-7
Merck Index: 14,3418

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR