Thaumatin (from Thaumatococcus daniellii), powder
Supplier: Thermo Scientific Chemicals
J63668-MC
J63668-MD
J63668-03
CAAAJ63668-MCEA
193.66
CAD
CAAAJ63668-MC
CAAAJ63668-MD
CAAAJ63668-03
Thaumatin (from Thaumatococcus daniellii), powder
Thaumatin
A mixture of the intensely sweet proteins thaumatin I and thaumatin II
Powder
Applications: A mixture of the intensely sweet proteins thaumatin I and thaumatin II
Storage Temperature:
Ambient |
MDL Number:
MFCD00132420 CAS Number: 53850-34-3 EINECS: 258-822-2 Merck Index: 14,9273 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...