Citrate-Buffered Saline (20X), Thermo Scientific
Supplier: Thermo Scientific Chemicals
J63666-K2
J63666-K3
CAAAJ63666-K2EA
51.07
CAD
CAAAJ63666-K2
CAAAJ63666-K3
Citrate-Buffered Saline (20X), Thermo Scientific
Buffers
Electrophoresis Buffers
Hybridisation Buffers
Saline-sodium citrate (SSC) buffer.
300 mM sodium citrate, 3M sodium chloride, pH 7.0