Clomiphene citrate salt ≥98%

Supplier: Thermo Scientific Chemicals
Danger

J63663-04 J63663-09
CAAAJ63663-04EA 186.55 CAD
CAAAJ63663-04 CAAAJ63663-09
Clomiphene citrate salt ≥98%
Clomiphene citrate salt

A selective estrogen receptor modulator


Crystalline powder


Applications: A selective estrogen receptor modulator

Formula: C₃₂H₃₆ClNO₈
Storage Temperature: Ambient
MDL Number: MFCD00058322
CAS Number: 50-41-9
EINECS: 200-035-3
Merck Index: 14,2386

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR