Temsirolimus ≥99%

Supplier: Thermo Scientific Chemicals

J63654-MB J63654-MC J63654-MF
CAAAJ63654-MBEA 689.42 CAD
CAAAJ63654-MB CAAAJ63654-MC CAAAJ63654-MF
Temsirolimus ≥99%
Temsirolimus

A Rapamycin derivative with anti-cancer activity that inhibits mTOR


Powder


Applications: A Rapamycin derivative with anti-cancer activity that inhibits mTOR

Formula: C₅₆H₈₇NO₁₆
Storage Temperature: Refrigerator
MDL Number: MFCD00934421
CAS Number: 162635-04-3
Merck Index: 14,9142

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR