Etoposide

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: (5R,5aR,8aR,9S)-9-(((4aR,6R,7R,8R,8aS)-7,8-Dihydroxy-2-methylhexahydropyrano[3,2-d][1,3]dioxin-6-yl)oxy)-5-(4-hydroxy-3,5-dimethoxyphenyl)-5,5a,8a,9-tetrahydrofuro[3',4':6,7]naphtho[2,3-d][1,3]dioxol-6(8H)-one, 4'-Demethylepipodophyllotoxin 9-(4,6-O-Ethylidene-beta-D-glucopyranoside)

J63651-MB J63651-MC
CAAAJ63651-MBEA 159.34 CAD
CAAAJ63651-MB CAAAJ63651-MC
Etoposide
Etoposide

Topoisomerase inhibitor and inducer of apoptosis


Powder


Applications: Topoisomerase inhibitor and inducer of apoptosis

Formula: C₂₉H₃₂O₁₃
Melting Pt: 236…251 °C
Storage Temperature: Ambient
MDL Number: MFCD00869325
CAS Number: 33419-42-0
EINECS: 251-509-1
Merck Index: 14,3886

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR