1-Aminobenzotriazole 97%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 1H-Benzo[d][1,2,3]triazol-1-amine

J63610-03 J63610-06 J63610-MD
CAAAJ63610-03EA 721.48 CAD
CAAAJ63610-03 CAAAJ63610-06 CAAAJ63610-MD
1-Aminobenzotriazole 97%
1-Aminobenzotriazole

Suicide inhibitor of cytochrome P450 and chloroperoxidase


Powder


Suicide inhibitor of cytochrome P450 and chloroperoxidase

Formula: C₆H₆N₄
MW: 134.14 g/mol
Melting Pt: 81…85 °C
Storage Temperature: Ambient
MDL Number: MFCD00132902
CAS Number: 1614-12-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR