Vinblastine sulfate 98%

Supplier: Thermo Scientific Chemicals
Danger

J63598-MA J63598-MF
CAAAJ63598-MAEA 263.88 CAD
CAAAJ63598-MA CAAAJ63598-MF
Vinblastine sulfate 98%
Vinblastine sulfate

Anticancer agent; microtubule disrupter; Induces apoptosis


Powder


Applications: Anticancer agent; microtubule disrupter; Induces apoptosis

Formula: C₄₆H₆₀N₄O₁₃S
Melting Pt: 267 °C
Storage Temperature: Ambient
MDL Number: MFCD00082457
CAS Number: 143-67-9
EINECS: 205-606-0
Merck Index: 14,9982

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR