Captopril

Supplier: Thermo Scientific Chemicals
Danger

J63593-03 J63593-06 J63593-14
CAAAJ63593-03EA 73.52 CAD
CAAAJ63593-03 CAAAJ63593-06 CAAAJ63593-14
Captopril
Captopril

Orally active inhibitor of angiotensin-converting enzyme


Powder


Applications: Orally active inhibitor of angiotensin-converting enzyme

Formula: C₉H₁₅NO₃S
Melting Pt: 104…108 °C
Storage Temperature: Ambient
MDL Number: MFCD00168073
CAS Number: 62571-86-2
EINECS: 263-607-1
Merck Index: 14,1774

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR