Thiamphenicol

Supplier: Thermo Scientific Chemicals

J63575-03 J63575-06 J63575-14
CAAAJ63575-03EA 64.76 CAD
CAAAJ63575-03 CAAAJ63575-06 CAAAJ63575-14
Thiamphenicol
Thiamphenicol

The methyl-sulfonyl analogue of chloramphenicol


Powder


Applications: The methyl-sulfonyl analogue of chloramphenicol

Formula: C₁₂H₁₅Cl₂NO₅S
Storage Temperature: Ambient
MDL Number: MFCD00467983
CAS Number: 15318-45-3
EINECS: 239-355-3
Merck Index: 14,9301

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR