N-Methyl-N-octanoyl-D-glucamine (MEGA-8)

Supplier: Thermo Scientific Chemicals

Synonyms: N-Octanoyl-N-methyl-D-glucamine, N-Methyl-N-((2S,3R,4R,5R)-2,3,4,5,6-pentahydroxyhexyl)octanamide, MEGA-8

J63574-03 J63574-06
CAAAJ63574-03EA 182.1 CAD
CAAAJ63574-03 CAAAJ63574-06
N-Methyl-N-octanoyl-D-glucamine (MEGA-8)
N-Methyl-N-octanoyl-D-glucamine (MEGA-8)

Non-ionic detergent for solubilizing membrane proteins.


Powder


Non-ionic detergent for solubilizing membrane proteins

Formula: C₁₅H₃₁NO₆
MW: 321.4
Melting Pt: 80…90 °C
Storage Temperature: Ambient
MDL Number: MFCD00134152
CAS Number: 85316-98-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR