Nalidixic acid sodium salt
Supplier: Thermo Scientific Chemicals
|
Danger
|
J63550-06
J63550-14
J63550-22
CAAAJ63550-06EA
82.87
CAD
CAAAJ63550-06
CAAAJ63550-14
CAAAJ63550-22
Nalidixic acid sodium salt
Nalidixic acid sodium salt
Inhibitor of bacterial DNA gyrase. An antibacterial agent.
Powder
Inhibitor of bacterial DNA gyrase. An antibacterial agent
Formula:
C₁₂H₁₁N₂NaO₃ Melting Pt: >280 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD00064376 CAS Number: 3374-05-8 EINECS: 222-159-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...