Cetirizine dihydrochloride ≥99%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
Cetirizine 2HCl, 2-(2-(4-((4-Chlorophenyl)(phenyl)methyl)piperazin-1-yl)ethoxy)acetic acid dihydrochloride
J63549-06
J63549-09
J63549-14
CAAAJ63549-06EA
391.22
CAD
CAAAJ63549-06
CAAAJ63549-09
CAAAJ63549-14
Cetirizine dihydrochloride ≥99%
Cetirizine dihydrochloride
Histamine H-1 receptor antagonist. Inhibits activation of eosinophils and neutrophils
Solid
Applications: Histamine H-1 receptor antagonist. Inhibits activation of eosinophils and neutrophils
Formula:
C₂₁H₂₅ClN₂O₃ MW: 461.81 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00941428 CAS Number: 83881-52-1 Merck Index: 14,2022 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...