MOBS, 0.2 M Buffer Solution, pH 7.5, Thermo Scientific
Supplier: Thermo Scientific Chemicals
J63519-AE
J63519-AK
CAAAJ63519-AEEA
175.14
CAD
CAAAJ63519-AE
CAAAJ63519-AK
MOBS, 0.2 M Buffer Solution, pH 7.5, Thermo Scientific
Buffers
Biological Buffers
Liquid
CAS No.: 115724-21-5
Physical Form: Liquid
MDL No.: MFCD00674916
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...