L(-)-Tyrosine cell culture reagent

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: L-Tyrosine, (S)-(-)-Tyrosine, Tyrosine, H-Tyr-OH

J63511-09 J63511-22 J63511-36 J63511-A1
CAAAJ63511-09EA 23.15 CAD
CAAAJ63511-09 CAAAJ63511-22 CAAAJ63511-36 CAAAJ63511-A1
L(-)-Tyrosine cell culture reagent
L(-)-Tyrosine

Powder


CAS No.: 60-18-4 Molecular Formula: C9H11NO3 Formula Weight: 181.19 Physical Form: Powder Appearance: White to pale-brown Melting Point: 300° Beilstein Reference: 392441 Merck Reference: 14,9839 Optical Rotation: -11° (c=4 in 1N HCl) MDL No.: MFCD00002606

Formula: C₉H₁₁NO₃
Boiling Pt: 351 °C (1013 hPa)
Melting Pt: >300 °C
Density: 1.41 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00002606
CAS Number: 60-18-4
EINECS: 200-460-4
Merck Index: 14,9839

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR