Tamoxifen 98%

Supplier: Thermo Scientific Chemicals
Danger

J63509-03 J63509-06 J63509-ME
CAAAJ63509-03EA 448.88 CAD
CAAAJ63509-03 CAAAJ63509-06 CAAAJ63509-ME
Tamoxifen 98%
Tamoxifen

Inhibitor of protein kinase C and induces apoptosis in cancer cells


It is also used in the treatment of infertility in women with anovulatory disorders as well as to prevent estrogen-related gynecomastia. It is utilized in the study on lipopolysaccharide (LPS)-induced microglial activation to know its effect.


Applications: Inhibitor of protein kinase C and induces apoptosis in cancer cells

Formula: C₂₆H₂₉NO
Storage Temperature: Refrigerator
MDL Number: MFCD00010454
CAS Number: 10540-29-1
EINECS: 234-118-0
Merck Index: 14,9048

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR