Imidazole, 0.5 M Buffer Solution, pH 7.5, Thermo Scientific
Supplier: Thermo Scientific Chemicals
J63496-AE
J63496-AK
CAAAJ63496-AEEA
175.14
CAD
CAAAJ63496-AE
CAAAJ63496-AK
Imidazole, 0.5 M Buffer Solution, pH 7.5, Thermo Scientific
Buffers
Biological Buffers
Liquid
CAS No.: 288-32-4
Storage Temperature: 2°C to 8°C
Physical Form: Liquid
MDL No.: MFCD00005183
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...