6-Cyano-7-nitroquinoxaline-2,3-dione 98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
J63472-MA
J63472-MB
J63472-MF
CAAAJ63472-MAEA
268.98
CAD
CAAAJ63472-MA
CAAAJ63472-MB
CAAAJ63472-MF
6-Cyano-7-nitroquinoxaline-2,3-dione 98%
6-Cyano-7-nitroquinoxaline-2,3-dione
Potent, selective kainate/quisqualate (non-NMDA) receptor antagonist
Solid
Potent, selective kainate/quisqualate (non-NMDA) receptor antagonist
Formula:
C₉H₄N₄O₄ Melting Pt: >300 °C Storage Temperature: Ambient |
MDL Number:
MFCD00069232 CAS Number: 115066-14-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...