Finasteride

Supplier: Thermo Scientific Chemicals
Danger

J63454-03 J63454-06 J63454-MC J63454-ME
CAAAJ63454-03EA 331.8 CAD
CAAAJ63454-03 CAAAJ63454-06 CAAAJ63454-MC CAAAJ63454-ME
Finasteride
Finasteride

Inhibitor of steroid 5-a-reductase


Powder


Applications: Inhibitor of steroid 5-a-reductase

Formula: C₂₃H₃₆N₂O₂
Storage Temperature: Refrigerator
MDL Number: MFCD00869737
CAS Number: 98319-26-7
Merck Index: 14,4082

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR