Bromhexine hydrochloride ≥98%

Supplier: Thermo Scientific Chemicals

Synonyms: 2-Amino-3,5-dibromo-N-cyclohexyl-N-methylbenzylamine Hydrochloride, 2,4-Dibromo-6-((cyclohexyl(methyl)amino)methyl)aniline hydrochloride, N-(2-Amino-3,5-dibromobenzyl)-N-methylcyclohexylamine Hydrochloride

J63451-22 J63451-36
CAAAJ63451-22EA 330.25 CAD
CAAAJ63451-22 CAAAJ63451-36
Bromhexine hydrochloride ≥98%
Bromhexine hydrochloride

A mucolytic agent


Powder


Applications: A mucolytic agent

Formula: C₁₄H₂₀Br₂N₂
MW: 412.59 g/mol
Melting Pt: 240…244 °C
Storage Temperature: Ambient
MDL Number: MFCD00056626
CAS Number: 611-75-6
EINECS: 210-280-8
Merck Index: 14,1391

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR