Diatrizoic acid
Supplier: Thermo Scientific Chemicals
Synonyms:
3,5-Di(acetamido)-2,4,6-triiodobenzoic Acid, 3,5-Bis(acetamido)-2,4,6-triiodobenzoic Acid, 3,5-Diacetamido-2,4,6-triiodobenzoic acid
J63444-14
J63444-22
CAAAJ63444-06EA
42.82
CAD
CAAAJ63444-06
CAAAJ63444-14
CAAAJ63444-22
Diatrizoic acid
Diatrizoic acid
A high-osmolality radiocontrast agent
Powder
Applications: A high-osmolality radiocontrast agent
Formula:
C₁₁H₉I₃N₂O₄ MW: 613.92 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00069960 CAS Number: 117-96-4 EINECS: 204-223-6 Merck Index: 14,2992 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...