Sodium-4-aminobenzoate 98%

Supplier: Thermo Scientific Chemicals

Synonyms: 4-Aminobenzoic Acid Sodium Salt, Sodium 4-aminobenzoate

J63428-14 J63428-22
CAAAJ63428-14EA 108.07 CAD
CAAAJ63428-14 CAAAJ63428-22
Sodium-4-aminobenzoate 98%
Sodium-4-aminobenzoate

Pharmaceutical intermediate


Powder


Applications: Pharmaceutical intermediate

Formula: C₇H₇NO₂
MW: 159.12 g/mol
Melting Pt: >300 °C
Storage Temperature: Ambient
MDL Number: MFCD00064395
CAS Number: 555-06-6
EINECS: 209-080-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR