Sodium glutamate
Supplier: Thermo Scientific Chemicals
J63424-09
J63424-14
CAAAJ63424-09EA
115.1
CAD
CAAAJ63424-09
CAAAJ63424-14
Sodium glutamate
Sodium glutamate
Powder
CAS No.: 142-47-2
Molecular Formula: C5H8NNaO4
Formula Weight: 169.11
Physical Form: Powder
Appearance: White to off white
MDL No.: MFCD00013074
Formula:
C₅H₈NNaO₄ Storage Temperature: Ambient |
MDL Number:
MFCD00013074 CAS Number: 142-47-2 EINECS: 205-538-1 Merck Index: 14,6254 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...