Oxytocin

Supplier: Thermo Scientific Chemicals

J63421-LB0 J63421-MB
CAAAJ63421-LB0EA 480.95 CAD
CAAAJ63421-LB0 CAAAJ63421-MB
Oxytocin
Proteins and Peptides

Peptide hormone that acts predominantly as a neurotransmitter; stimulates uterine contraction and lactation.


  • EINECS: 200-048-4
  • GHS symbols and signal word: GHS06, GHS08 and Danger
  • Hazard class and packing group: 6.1, II
  • UN number: 2811
  • Merck index number: 14,6979


Purity: 96% CAS No.: 50-56-6 Molecular Formula: C43H66N12O12S2 Formula Weight: 1007.19 Storage Temperature: -30°C to -10°C Physical Form: Lyophilized powder Merck Reference: 14,6979 UN No.: 2811 MDL No.: MFCD00076731


Peptide hormone that acts predominantly as a neurotransmitter; stimulates uterine contraction and lactation

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR