Bisindolylmaleimide I

Supplier: Thermo Scientific Chemicals

Synonyms: 3-(1-(3-(Dimethylamino)propyl)-1H-indol-3-yl)-4-(1H-indol-3-yl)-1H-pyrrole-2,5-dione, 3-[1-[3-(Dimethylamino)propyl]-1H-indol-3-yl]-4-(1H-indol-3-yl)-1H-pyrrole-2,5-dione, GF 109203X

J63401-LB0 J63401-MB
CAAAJ63401-LB0EA 397.61 CAD
CAAAJ63401-LB0 CAAAJ63401-MB
Bisindolylmaleimide I
Bisindolylmaleimide I

Very potent and selective inhibitor of protein kinase C


Orange crystalline powder


Very potent and selective inhibitor of protein kinase C

Formula: C₂₅H₂₄N₄O₂
Storage Temperature: Refrigerator
MDL Number: MFCD00236428
CAS Number: 133052-90-1

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR