Geldanamycin ≥99%

Supplier: Thermo Scientific Chemicals

J63397-MB J63397-MC J63397-MD
CAAAJ63397-MBEA 452.13 CAD
CAAAJ63397-MB CAAAJ63397-MC CAAAJ63397-MD
Geldanamycin ≥99%
Geldanamycin

Benzoquinoid antibiotic that inhibits pp60src tyrosine kinase


Powder


Applications: Benzoquinoid antibiotic that inhibits pp60src tyrosine kinase

Formula: C₂₉H₄₀N₂O₉
Storage Temperature: Refrigerator
MDL Number: MFCD00274570
CAS Number: 30562-34-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR