Sodium dodecyl sulfate (SDS) 20% in aqueous solution

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: Sodium lauryl sulfate, Dodecyl sulfate sodium salt, Lauryl sulfate sodium salt, Dodecyl sulfuric acid sodium salt, Dodecyl sulphate sodium salt, Sodium dodecyl sulfate, Dodecyl sulphuric acid sodium salt, Lauryl sulphate sodium salt, Sodium lauryl sulphate

J63394-AK J63394-AP
CAAAJ63394-AKEA 125.07 CAD
CAAAJ63394-AK CAAAJ63394-AP
Sodium dodecyl sulfate (SDS) 20% in aqueous solution
Sodium dodecyl sulphate (SDS)

Liquid


CAS No.: 151-21-3 Molecular Formula: C12H25NaO4S Formula Weight: 288.38 Physical Form: Liquid Appearance: Colorless

Formula: CH₃(CH₂)₁₁OSO₃Na
MW: 288.38 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00036175
CAS Number: 151-21-3
EINECS: 205-788-1
Merck Index: 14,08636
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=fe4cb1741fba41f699a3be006d0b01b1b551898c&vid=c6df9a86fc351b319e68cc79c1c17f733b4fec67&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR