Cytidine-5'-monophosphate disodium salt ≥99%

Supplier: Thermo Scientific Chemicals

J63376-03 J63376-06 J63376-14
CAAAJ63376-03EA 72.13 CAD
CAAAJ63376-03 CAAAJ63376-06 CAAAJ63376-14
Cytidine-5'-monophosphate disodium salt ≥99%
Cytidine-5'-monophosphate disodium salt

Crystalline powder


Purity: 99+% CAS No.: 6757-06-8 Molecular Formula: C9H12N3Na2O8P Formula Weight: 367.16 Storage Temperature: 2°C to 8°C Physical Form: Crystalline powder Appearance: White MDL No.: MFCD00064355

Formula: C₉H₁₂N₃Na₂O₈P
Storage Temperature: Ambient
MDL Number: MFCD00064355
CAS Number: 6757-06-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR