Potassium acetate 8 M in aqueous solution

Supplier: Thermo Scientific Chemicals

J63372-AE J63372-AK
CAAAJ63372-AEEA 105.23 CAD
CAAAJ63372-AE CAAAJ63372-AK
Potassium acetate 8 M in aqueous solution
Potassium acetate

Liquid


CAS No.: 127-08-2 Molecular Formula: C2H3KO2 Formula Weight: 98.15 Physical Form: Liquid MDL No.: MFCD00012458

Formula: CH₃COOK
Storage Temperature: Ambient
MDL Number: MFCD00012458
CAS Number: 127-08-2
EINECS: 204-822-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=13cc4439b794fe60df11a635ce6d3a03e9609099&vid=06c778a684c28fe03ba0262abdbaa81c094e79a6&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR