Phosphate, 0.5 M Buffer Solution, pH 7.5, Thermo Scientific
Supplier: Thermo Scientific Chemicals
J63349-AK
J63349-AP
CAAAJ63349-AKEA
100.82
CAD
CAAAJ63349-AK
CAAAJ63349-AP
Phosphate, 0.5 M Buffer Solution, pH 7.5, Thermo Scientific
Buffers
Biological Buffers
Liquid
CAS No.: 7558-80-7
Physical Form: Liquid
MDL No.: MFCD00003527
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...