Tween 20 washing buffer, 1% in PBS (10X), Thermo Scientific

Supplier: Thermo Scientific Chemicals

J63314-AP J63314-K2 J63314-K3
CAAAJ63314-APEA 125.07 CAD
CAAAJ63314-AP CAAAJ63314-K2 CAAAJ63314-K3
Tween 20 washing buffer, 1% in PBS (10X), Thermo Scientific
Buffers Electrophoresis Buffers Washing Buffers

Viscous liquid


Storage Temperature: 2°C to 8°C
Physical Form: Viscous liquid

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR