Nicergoline ≥95%

Supplier: Thermo Scientific Chemicals
Warning

J63295-03 J63295-MC
CAAAJ63295-03EA 740.66 CAD
CAAAJ63295-03 CAAAJ63295-MC
Nicergoline ≥95%
Nicergoline

Cerebral and peripheral vasodilator


Powder


Applications: Cerebral and peripheral vasodilator

Formula: C₂₄H₂₆BrN₃O₃
Storage Temperature: Ambient
MDL Number: MFCD00869626
CAS Number: 27848-84-6
EINECS: 248-694-6
Merck Index: 14,6496

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR