Forskolin 98+%

Supplier: Thermo Scientific Chemicals
Warning

J63292-MA J63292-MF
CAAAJ63292-MAEA 452.13 CAD
CAAAJ63292-MA CAAAJ63292-MF
Forskolin 98+%
Forskolin

An adenylcyclase activator


Solid


An adenylcyclase activator

Formula: C₂₂H₃₄O₇
Storage Temperature: Refrigerator
MDL Number: MFCD00082317
CAS Number: 66575-29-9
EINECS: 266-410-9
Merck Index: 14,2476

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR