Fexofenadine hydrochloride

Supplier: Thermo Scientific Chemicals

Synonyms: 2-(4-(1-Hydroxy-4-(4-(hydroxydiphenylmethyl)piperidin-1-yl)butyl)phenyl)-2-methylpropanoic acid hydrochloride, Fexofenadine HCl

J63262-MB J63262-MC
CAAAJ63262-MBEA 251.58 CAD
CAAAJ63262-MB CAAAJ63262-MC CAAAJ63262-ME
Fexofenadine hydrochloride
Fexofenadine hydrochloride

H-1 receptor antagonist


Crystalline powder


Applications: H-1 receptor antagonist

Formula: C₃₂H₃₉NO₄
MW: 538.13 g/mol
Melting Pt: 148…150 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00865710
CAS Number: 153439-40-8
Merck Index: 14,4068

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR