MES, 0.2 M Buffer Solution, pH 5.5, Thermo Scientific

Supplier: Thermo Scientific Chemicals

J63257-AK J63257-AP
CAAAJ63257-AKEA 116.51 CAD
CAAAJ63257-AK CAAAJ63257-AP
MES, 0.2 M Buffer Solution, pH 5.5, Thermo Scientific
Buffers Biological Buffers

Liquid


CAS No.: 145224-94-8
Physical Form: Liquid
Appearance: Colorless
MDL No.: MFCD00283848

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR