Bifonazole 99%

Supplier: Thermo Scientific Chemicals
Warning

J63253-03 J63253-06
CAAAJ63253-03EA 131.42 CAD
CAAAJ63253-03 CAAAJ63253-06 CAAAJ63253-14
Bifonazole 99%
Bifonazole

Inhibits synthesis of ergosterol in dermatophytes. Calmodulin antagonist


Powder


Applications: Inhibits synthesis of ergosterol in dermatophytes. Calmodulin antagonist

Formula: C₂₂H₁₈N₂
Melting Pt: 142 °C
Storage Temperature: Ambient
MDL Number: MFCD00865567
CAS Number: 60628-96-8
EINECS: 262-336-6
Merck Index: 14,1213

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR