(+)-cis-Diltiazem hydrochloride
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
(2S,3S)-(+)-3-Acetoxy-2,3-dihydro-5-[2-(dimethylamino)ethyl]-2-(4-methoxyphenyl)-1,5-benzothiazepin-4(5H)-one Hydrochloride, Diltiazem Hydrochloride, (+)-cis-Diltiazem HCl, (+)-cis-3-Acetoxy-5-(2-dimethylaminoethyl)-2,3-dihydro-2-(4-methoxyphenyl)-1,5-benzothiazepin-4(5H)-one Hydrochloride, (2S,3S)-5-(2-(Dimethylamino)ethyl)-2-(4-methoxyphenyl)-4-oxo-2,3,4,5-tetrahydrobenzo[b][1,4]thiazepin-3-yl acetate hydrochloride
An L-type calcium channel blocker
Powder
Applications: An L-type calcium channel blocker
Formula:
C₂₂H₂₆N₂O₄S·HCl Melting Pt: 207…212 °C Storage Temperature: Ambient |
MDL Number:
MFCD00069252 CAS Number: 33286-22-5 EINECS: 251-443-3 Merck Index: 14,3202 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...