L(-)-Malic acid 99%
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
L(-)-Malic acid, (S)-(-)-Hydroxy-1,4-butanedioic acid, (S)-(-)-Hydroxysuccinic acid, (S)-2-hydroxysuccinic acid, L(-)-Hydroxysuccinic acid, L(-)-Hydroxy-1,4-butanedioic acid, L-(–)-Malic Acid
Crystalline powder
Purity: 99%
CAS No.: 97-67-6
Molecular Formula: C4H6O5
Formula Weight: 134.09
Flash Point: 220°(428°F)
Physical Form: Crystalline powder
Density: 1.6
Appearance: White
Melting Point: 100-106°
Beilstein Reference: 1723541
Merck Reference: 14,5707
Optical Rotation: -26° (c=5.5 in pyridine)
MDL No.: MFCD00064213
Formula:
C₄H₆O₅ MW: 134.09 g/mol Boiling Pt: 322 Melting Pt: 100…106 °C Density: 1.6 Flash Pt: 220 °C(428 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00064213 CAS Number: 97-67-6 EINECS: 202-601-5 Merck Index: 14,5707 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...