Urapidil hydrochloride ≥98%

Supplier: Thermo Scientific Chemicals
Warning

J63219-03 J63219-ME
CAAAJ63219-03EA 565.97 CAD
CAAAJ63219-03 CAAAJ63219-06 CAAAJ63219-ME
Urapidil hydrochloride ≥98%
Urapidil hydrochloride

alpha-1-Adrenoceptor antagonist and 5-HT1A receptor agonist


Powder


Applications: alpha-1-Adrenoceptor antagonist and 5-HT1A receptor agonist

Formula: C₂₀H₂₉N₅O₃·HCl
Storage Temperature: Ambient
MDL Number: MFCD00078601
CAS Number: 64887-14-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR