N-(1-Naphthyl)ethylenediamine dihydrochloride ACS
Supplier: Thermo Scientific Chemicals
|
Warning
|
J63214-06
J63214-14
J63214-22
CAAAJ63214-06EA
66.2
CAD
CAAAJ63214-06
CAAAJ63214-14
CAAAJ63214-22
N-(1-Naphthyl)ethylenediamine dihydrochloride ACS
N-2-Aminoethyl-1-naphthylamine dihydrochloride
Used for colorimetric determination of sulfonamide in blood.
Powder
Used for colorimetric determination of sulfonamide in blood
Formula:
C₁₂H₁₆Cl₂N₂ Melting Pt: 191…199 °C Storage Temperature: Ambient |
MDL Number:
MFCD00012556 CAS Number: 1465-25-4 EINECS: 215-981-2 Merck Index: 14,6409 |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...