Galactitol 99%

Supplier: Thermo Scientific Chemicals

Synonyms: Dulcitol, rel-(2R,3S,4R,5S)-Hexane-1,2,3,4,5,6-hexaol

J63206-30 J63206-36
CAAAJ63206-30EA 460.11 CAD
CAAAJ63206-30 CAAAJ63206-36
Galactitol 99%
Galactitol

Useful reagent in bacteriology


Powder


Useful reagent in bacteriology

Formula: HOH₂C(CH(OH))₄CH₂OH
Boiling Pt: 275…280 °C/1 mm
Melting Pt: 187…191 °C
Density: 1.47
Storage Temperature: Ambient
MDL Number: MFCD00064288
CAS Number: 608-66-2
EINECS: 210-165-2
Merck Index: 14,4332

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR