Econazole Nitrate
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
1-[2-(4-Chlorobenzyloxy)-2-(2,4-dichlorophenyl)ethyl]imidazole Nitrate, Ecostatin, 1-[2-(4-Chlorophenylmethoxy)-2-(2,4-dichlorophenyl)ethyl]-1H-imidazole nitrate
J63173-06
J63173-14
CAAAJ63173-06EA
155.55
CAD
CAAAJ63173-06
CAAAJ63173-14
Econazole Nitrate
Econazole nitrate
Powder
CAS No.: 24169-02-6
Molecular Formula: C18H15Cl3N2O.HNO3
Formula Weight: 444.70
Physical Form: Powder
Appearance: White to off-white
Melting Point: 158-163°
Solubility: Soluble at 0.4mg/ml in DMSO. Sparingly soluble in water.
MDL No.: MFCD00058160
Formula:
C₁₈H₁₅Cl₃N₂O·HNO₃ Melting Pt: 158…163 °C Storage Temperature: Ambient |
MDL Number:
MFCD00058160 CAS Number: 24169-02-6 EINECS: 246-053-5 Merck Index: 14,3502 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...