Naringin

Supplier: Thermo Scientific Chemicals

J63166-14 J63166-22
CAAAJ63166-14EA 86.62 CAD
CAAAJ63166-14 CAAAJ63166-22
Naringin
Naringin

A citrus bioflavanoid that inhibits cytochrome P-450 monooxygenase activity


Powder


Applications: A citrus bioflavanoid that inhibits cytochrome P-450 monooxygenase activity

Formula: C₂₇H₃₂O₁₄
Storage Temperature: Ambient
MDL Number: MFCD00148888
CAS Number: 10236-47-2
EINECS: 233-566-4
Merck Index: 14,6425

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR