β-Cyclodextrine

Supplier: Thermo Scientific Chemicals

Synonyms: Caraway, Cycloheptaamylose, β-Cyclodextrin, Cyclomaltoheptaose, Schardinger β-Dextrin, Betadex

J63161-14 J63161-22
CAAAJ63161-14EA 72.13 CAD
CAAAJ63161-14 CAAAJ63161-22
β-Cyclodextrine
β-Cyclodextrine

Powder


Use to solubilize non-polar compounds such a fatty acids, lipids and cholesterol. Reported useful for the selective precipitation of enantiomeric, positional or structural isomers


Use to solubilize non-polar compounds such a fatty acids, lipids and cholesterol. Reported useful for the selective precipitation of enantiomeric, positional or structural isomers

Formula: C₄₂H₇₀O₃₅
MW: 1134.98 g/mol
Melting Pt: 290 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00078139
CAS Number: 7585-39-9
EINECS: 231-493-2
Merck Index: 14,2718

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR