α-Ketoglutaric acid disodium salt dihydrate 99%
Supplier: Thermo Scientific Chemicals
J63154-14
J63154-22
J63154-30
J63154-03
CAAAJ63154-14EA
119.91
CAD
CAAAJ63154-14
CAAAJ63154-22
CAAAJ63154-30
CAAAJ63154-03
α-Ketoglutaric acid disodium salt dihydrate 99%
α-Ketoglutaric acid disodium salt dihydrate
Powder
Purity: 99%
CAS No.: 305-72-6
Molecular Formula: C5H4Na2O5.2H2O
Formula Weight: 226.09 (190.06anhy)
Physical Form: Powder
Sensitivity: Hygroscopic
Appearance: White
MDL No.: MFCD00150702
Formula:
C₅H₄O₅·2Na·2H₂O Storage Temperature: Ambient |
MDL Number:
MFCD00150702 CAS Number: 1282616-74-3 EINECS: 206-167-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...