Manganese(II) chloride 1 M in aqueous solution

Supplier: Thermo Scientific Chemicals
Danger

J63150-AD J63150-AE
CAAAJ63150-ADEA 97.04 CAD
CAAAJ63150-AD CAAAJ63150-AE
Manganese(II) chloride 1 M in aqueous solution
Manganese (II) chloride

Liquid


CAS No.: 7773-01-5 Molecular Formula: Cl2Mn Formula Weight: 125.84 Physical Form: Liquid Appearance: Colorless

Formula: MnCl₂
Storage Temperature: Ambient
MDL Number: MFCD00011114
CAS Number: 7773-01-5
EINECS: 231-869-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR