2-Amino-2-methyl-1,3-propanediol 99+%

Supplier: Thermo Scientific Chemicals
Warning

J63144-14 J63144-22 J63144-30
CAAAJ63144-14EA 51.46 CAD
CAAAJ63144-14 CAAAJ63144-22 CAAAJ63144-30
2-Amino-2-methyl-1,3-propanediol 99+%
2-Amino-2-methyl-1,3-propanediol

A useful biological buffer.


Powder


A useful biological buffer

Formula: C₄H₁₁NO₂
Boiling Pt: 151 °C/10 mm
Melting Pt: 100…110 °C
Density: 1.211 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00004678
CAS Number: 115-69-5
EINECS: 204-100-7
UN: 3259
ADR: 8,III
Merck Index: 14,448

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR